Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
spop-1
Entrez Gene ID
176422
Gene Name
protein-coding
Gene Aliases
CELE_T16H12.5
Chromosome Location
Chr.III: 10090862 - 10098081 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027593.2 NP_001022764.1 1 - 2 93 73

View Full Product Details