Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mua-3
Entrez Gene ID
176430
Gene Name
protein-coding
Gene Aliases
CELE_K08E5.3
Chromosome Location
Chr.III: 10160697 - 10183529 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027503.4 NP_001022674.1 27 - 28 5883 80
NM_001027504.3 NP_001022675.1 17 - 18 4523 80
GenBank mRNA AF139060.1 - 27 - 28 5883 80

View Full Product Details