Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
chc-1
Entrez Gene ID
176434
Gene Name
Probable clathrin heavy chain 1
Gene Aliases
CELE_T20G5.1
Chromosome Location
Chr.III: 10203821 - 10209704 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066859.5 NP_499260.1 9 - 10 4961 107

View Full Product Details