Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cts-1
Entrez Gene ID
176437
Gene Name
Probable citrate synthase, mitochondrial
Gene Aliases
CELE_T20G5.2
Chromosome Location
Chr.III: 10214771 - 10216635 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066863.9 NP_499264.1 2 - 3 621 68

View Full Product Details