Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
div-1
Entrez Gene ID
176439
Gene Name
DNA polymerase alpha subunit B
Gene Aliases
CELE_R01H10.1
Chromosome Location
Chr.III: 10245056 - 10248905 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066867.5 NP_499268.1 3 - 4 965 132

View Full Product Details