Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bbs-5
Entrez Gene ID
187516
Gene Name
Bardet-Biedl syndrome 5 protein homolog
Gene Aliases
CELE_R01H10.6
Chromosome Location
Chr.III: 10258831 - 10261782 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066871.2 NP_499272.1 1 - 2 191 64

View Full Product Details