Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ced-9
- Entrez Gene ID
-
3565776
- Gene Name
- Apoptosis regulator ced-9
- Gene Aliases
-
CELE_T07C4.8
- Chromosome Location
-
Chr.III: 10335172 - 10337542 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066883.6 |
NP_499284.1 |
3 - 4 |
666 |
81 |
- Gene Symbol
- mev-1
- Entrez Gene ID
-
260040
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_T07C4.7
- Chromosome Location
-
Chr.III: 10334277 - 10337542 on Build WBcel235
View Full Product Details