Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbc-8
Entrez Gene ID
176454
Gene Name
protein-coding
Gene Aliases
CELE_C38H2.1
Chromosome Location
Chr.III: 10364951 - 10373234 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268143.3 NP_001255072.1 7 - 8 1428 73
NM_001268144.3 NP_001255073.1 3 - 4 629 73

View Full Product Details