Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cul-1
Entrez Gene ID
176466
Gene Name
Cullin-1
Gene Aliases
CELE_D2045.6
Chromosome Location
Chr.III: 10469716 - 10475245 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066908.7 NP_499309.1 3 - 4 1399 99
GenBank mRNA U58083.1 - 2 - 3 1375 99

View Full Product Details