Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab-33
Entrez Gene ID
176470
Gene Name
Ras-related protein Rab-33
Gene Aliases
CELE_F43D9.2
Chromosome Location
Chr.III: 10500209 - 10503853 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066913.6 NP_499314.1 3 - 4 557 114

View Full Product Details