The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001027611.3 | NP_001022782.1 | 8 - 9 | 1065 | 135 |
NM_001379903.1 | NP_001368253.1 | 4 - 5 | 438 | 135 | |
GenBank mRNA | AF151640.1 | - | 8 - 9 | 1234 | 135 |