Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rabx-5
Entrez Gene ID
176481
Gene Name
protein-coding
Gene Aliases
CELE_Y39A1A.5
Chromosome Location
Chr.III: 10605343 - 10614930 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372654.2 NP_001360492.1 7 - 8 1275 81
NM_001382984.1 7 - 8 1293 81

Gene Symbol
Y39A1A.24
Entrez Gene ID
3564902
Gene Name
Uncharacterized protein
Gene Aliases
CELE_Y39A1A.24
Chromosome Location
Chr.III: 10605342 - 10614935 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379906.2 NP_001366657.1 7 - 8 1303 81

View Full Product Details