Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhs-11
Entrez Gene ID
176485
Gene Name
protein-coding
Gene Aliases
CELE_Y39A1A.11
Chromosome Location
Chr.III: 10640828 - 10642865 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066945.3 NP_499346.2 3 - 4 288 71

View Full Product Details