Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- hex-3
- Entrez Gene ID
-
189716
- Gene Name
- Glyco_hydro_20 domain-containing protein
- Gene Aliases
-
CELE_Y39A1C.4
- Chromosome Location
-
Chr.III: 10849222 - 10855924 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066989.6 |
NP_499390.3 |
4 - 5 |
1559 |
91 |
- Gene Symbol
- Y39A1C.1
- Entrez Gene ID
-
189715
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_Y39A1C.1
- Chromosome Location
-
Chr.III: 10849224 - 10858174 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001382992.1 |
|
4 - 5 |
1557 |
91 |
View Full Product Details