Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
athp-1
Entrez Gene ID
176517
Gene Name
AT Hook plus PHD finger transcription factor
Gene Aliases
CELE_C44B9.4
Chromosome Location
Chr.III: 10881766 - 10893696 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066996.6 NP_499397.2 2 - 3 277 74

View Full Product Details