Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vab-7
Entrez Gene ID
176521
Gene Name
Homeobox protein vab-7
Gene Aliases
CELE_M142.4
Chromosome Location
Chr.III: 10920336 - 10926041 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067000.5 NP_499401.1 2 - 3 233 61

View Full Product Details