Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbx-34
Entrez Gene ID
176549
Gene Name
Putative T-box protein 34
Gene Aliases
CELE_Y47D3A.10
Chromosome Location
Chr.III: 11216398 - 11218277 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067040.5 NP_499441.1 2 - 3 387 81

View Full Product Details