Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sbp-1
Entrez Gene ID
176574
Gene Name
BHLH domain-containing protein
Gene Aliases
CELE_Y47D3B.7
Chromosome Location
Chr.III: 11428367 - 11442572 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067071.7 NP_499472.1 2 - 3 219 67

View Full Product Details