Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bed-2
Entrez Gene ID
176576
Gene Name
protein-coding
Gene Aliases
CELE_Y47D3B.9
Chromosome Location
Chr.III: 11460030 - 11470823 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001374881.2 NP_001362179.1 6 - 6 1515 85
NM_001374882.3 NP_001362180.1 4 - 4 1403 85

View Full Product Details