Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gly-8
Entrez Gene ID
176595
Gene Name
Probable N-acetylgalactosaminyltransferase 8
Gene Aliases
CELE_Y66A7A.6
Chromosome Location
Chr.III: 11624988 - 11628239 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379936.2 NP_001366981.1 7 - 8 912 123
GenBank mRNA AF031842.1 - 6 - 7 836 123

View Full Product Details