Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
maa-1
Entrez Gene ID
176612
Gene Name
ACB domain-containing protein
Gene Aliases
CELE_C18D11.2
Chromosome Location
Chr.III: 11819743 - 11822526 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067130.5 NP_499531.1 2 - 3 551 67

View Full Product Details