Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zyg-8
Entrez Gene ID
176639
Gene Name
protein-coding
Gene Aliases
CELE_Y79H2A.11
Chromosome Location
Chr.III: 12058836 - 12074168 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001347293.3 NP_001334209.1 2 - 3 867 74
NM_067170.7 NP_499571.2 4 - 5 1411 74
GenBank mRNA AJ319870.1 - 3 - 4 1326 74

View Full Product Details