Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zyg-8
Entrez Gene ID
176639
Gene Name
protein-coding
Gene Aliases
CELE_Y79H2A.11
Chromosome Location
Chr.III: 12058836 - 12074168 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001347293.3 NP_001334209.1 7 - 8 1773 70
NM_067170.7 NP_499571.2 9 - 10 2317 70
GenBank mRNA AJ319870.1 - 8 - 9 2232 70

View Full Product Details