Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mrp-8
Entrez Gene ID
190710
Gene Name
Uncharacterized protein
Gene Aliases
CELE_Y75B8A.26
Chromosome Location
Chr.III: 12283133 - 12303216 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067197.4 NP_499598.2 12 - 13 1795 75

View Full Product Details