Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-92
Entrez Gene ID
190711
Gene Name
Prion-like-(Q/N-rich)-domain-bearing protein
Gene Aliases
CELE_Y75B8A.27
Chromosome Location
Chr.III: 12307781 - 12311925 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067198.5 NP_499599.2 2 - 3 512 84

View Full Product Details