Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- pqn-92
- Entrez Gene ID
-
190711
- Gene Name
- Prion-like-(Q/N-rich)-domain-bearing protein
- Gene Aliases
-
CELE_Y75B8A.27
- Chromosome Location
-
Chr.III: 12307781 - 12311925 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067198.5 |
NP_499599.2 |
3 - 4 |
621 |
70 |
View Full Product Details