Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rsp-3
Entrez Gene ID
176688
Gene Name
Probable splicing factor, arginine/serine-rich 3
Gene Aliases
CELE_Y111B2A.18
Chromosome Location
Chr.III: 12686701 - 12687916 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383015.1 1 - 2 180 83

View Full Product Details