Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ssl-1
Entrez Gene ID
190954
Gene Name
protein-coding
Gene Aliases
CELE_Y111B2A.22
Chromosome Location
Chr.III: 12714566 - 12744721 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268250.2 NP_001255179.1 9 - 10 4406 93
NM_001268251.3 NP_001255180.1 9 - 10 4384 93
NM_001379967.1 NP_001367266.1 7 - 8 3946 93
GenBank mRNA AY551965.1 - 9 - 10 4428 93

View Full Product Details