Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arx-5
Entrez Gene ID
176697
Gene Name
Probable actin-related protein 2/3 complex subunit 3
Gene Aliases
CELE_Y37D8A.1
Chromosome Location
Chr.III: 12816789 - 12818100 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067266.4 NP_499667.1 1 - 2 116 92

View Full Product Details