Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-25
Entrez Gene ID
176713
Gene Name
protein-coding
Gene Aliases
CELE_Y37D8A.23
Chromosome Location
Chr.III: 12942333 - 12949884 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027661.5 NP_001022832.1 4 - 5 1076 137
NM_067288.5 NP_499689.1 6 - 7 1309 137
GenBank mRNA AF109378.1 - 6 - 7 1271 137

View Full Product Details