Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snf-7
Entrez Gene ID
176723
Gene Name
Transporter
Gene Aliases
CELE_ZK1010.9
Chromosome Location
Chr.III: 13005461 - 13011155 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067301.4 NP_499702.2 7 - 8 2033 116

View Full Product Details