Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zig-8
Entrez Gene ID
176732
Gene Name
Zwei Ig domain protein zig-8
Gene Aliases
CELE_Y39E4B.8
Chromosome Location
Chr.III: 13119820 - 13133938 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067313.7 NP_499714.1 1 - 2 92 79
GenBank mRNA AF456253.1 - 1 - 2 53 79

View Full Product Details