Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
abce-1
Entrez Gene ID
176733
Gene Name
ABC transporter, class E
Gene Aliases
CELE_Y39E4B.1
Chromosome Location
Chr.III: 13153662 - 13160288 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067316.7 NP_499717.1 4 - 5 1436 77

View Full Product Details