Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tsp-3
Entrez Gene ID
192059
Gene Name
TetraSPanin family
Gene Aliases
CELE_Y39E4B.4
Chromosome Location
Chr.III: 13212666 - 13214463 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067323.3 NP_499724.1 4 - 5 499 79

View Full Product Details