Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
klp-19
Entrez Gene ID
176750
Gene Name
Kinesin motor domain-containing protein
Gene Aliases
CELE_Y43F4B.6
Chromosome Location
Chr.III: 13302717 - 13307259 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067341.4 NP_499742.1 6 - 7 3067 85

View Full Product Details