Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kel-3
Entrez Gene ID
176775
Gene Name
protein-coding
Gene Aliases
CELE_T27E9.4
Chromosome Location
Chr.III: 13464820 - 13471350 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067383.6 NP_499784.2 2 - 3 300 87
NM_067384.6 NP_499785.2 2 - 3 291 87

View Full Product Details