Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acc-4
Entrez Gene ID
176778
Gene Name
Acetylcholine-gated ion channel acc-4
Gene Aliases
CELE_T27E9.9
Chromosome Location
Chr.III: 13480670 - 13484162 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383026.1 8 - 9 1112 73

View Full Product Details