Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-40
Entrez Gene ID
189006
Gene Name
protein-coding
Gene Aliases
CELE_T28A8.1
Chromosome Location
Chr.III: 13486514 - 13492477 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268276.2 NP_001255205.1 4 - 5 650 117
NM_001268277.1 NP_001255206.1 4 - 5 666 117
NM_001268278.2 NP_001255207.1 3 - 4 537 117

View Full Product Details