Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mlh-1
Entrez Gene ID
176783
Gene Name
DNA_mis_repair domain-containing protein
Gene Aliases
CELE_T28A8.7
Chromosome Location
Chr.III: 13509297 - 13514570 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067395.6 NP_499796.2 2 - 3 398 61

View Full Product Details