Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
osm-7
Entrez Gene ID
176790
Gene Name
protein-coding
Gene Aliases
CELE_T05D4.4
Chromosome Location
Chr.III: 13576188 - 13580662 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268280.2 NP_001255209.1 9 - 10 1387 74

View Full Product Details