Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acbp-5
Entrez Gene ID
176800
Gene Name
Acyl-Coenzyme A Binding Protein
Gene Aliases
CELE_T12D8.3
Chromosome Location
Chr.III: 13639891 - 13641688 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067416.6 NP_499817.2 2 - 3 413 88

View Full Product Details