Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dyf-2
Entrez Gene ID
191342
Gene Name
protein-coding
Gene Aliases
CELE_ZK520.3
Chromosome Location
Chr.III: 13674879 - 13686445 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083186.4 NP_001076655.1 7 - 8 1366 78
GenBank mRNA DQ314286.1 - 7 - 8 1366 78

View Full Product Details