Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cul-2
Entrez Gene ID
176806
Gene Name
protein-coding
Gene Aliases
CELE_ZK520.4
Chromosome Location
Chr.III: 13687263 - 13692683 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379988.1 NP_001367020.1 6 - 7 2019 87

View Full Product Details