Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lit-1
Entrez Gene ID
176808
Gene Name
protein-coding
Gene Aliases
CELE_W06F12.1
Chromosome Location
Chr.III: 13701986 - 13724404 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027634.3 NP_001022805.1 4 - 5 534 67
NM_001027637.5 NP_001022808.1 5 - 6 566 67
GenBank mRNA AF143243.1 - 3 - 4 377 67
AF145376.1 - 4 - 5 534 67

View Full Product Details