Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tyr-2
Entrez Gene ID
176810
Gene Name
TYRosinase
Gene Aliases
CELE_K08E3.1
Chromosome Location
Chr.III: 13744222 - 13747390 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067435.7 NP_499836.1 3 - 4 531 70

View Full Product Details