Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scl-23
Entrez Gene ID
182028
Gene Name
SCP domain-containing protein
Gene Aliases
CELE_B0545.3
Chromosome Location
Chr.IV: 86840 - 90091 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067458.5 NP_499859.3 6 - 7 806 63

View Full Product Details