Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tpa-1
Entrez Gene ID
176825
Gene Name
protein-coding
Gene Aliases
CELE_B0545.1
Chromosome Location
Chr.IV: 76328 - 110819 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372683.1 NP_001359808.1 5 - 6 568 66
NM_001372684.2 NP_001359809.1 5 - 6 484 66

View Full Product Details