Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-99
Entrez Gene ID
185112
Gene Name
protein-coding
Gene Aliases
CELE_F29C4.8
Chromosome Location
Chr.IV: 137974 - 147919 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129303.4 NP_001122775.2 14 - 15 1664 55
NM_001307632.3 NP_001294561.1 12 - 13 1601 55
NM_001307633.3 NP_001294562.1 12 - 13 1565 55
NM_001307634.4 NP_001294563.1 13 - 14 1637 55
NM_001322764.3 NP_001309612.1 11 - 12 1529 55
NM_001322765.3 NP_001309608.1 11 - 12 1529 55
NM_001322766.3 NP_001309610.1 12 - 13 1601 55
NM_001322767.3 NP_001309606.1 12 - 13 1601 55
NM_001322768.3 NP_001309611.1 12 - 13 1565 55
NM_001322769.3 NP_001309607.1 12 - 13 1565 55
NM_001322770.3 NP_001309609.1 13 - 14 1637 55
NM_001322771.3 NP_001309605.1 13 - 14 1637 55
NM_001322772.3 NP_001309604.1 11 - 12 1529 55
NM_001322773.3 NP_001309602.1 11 - 12 1529 55
NM_001322774.3 NP_001309603.1 12 - 13 1601 55
NM_001322775.2 NP_001309601.1 12 - 13 1565 55
NM_067468.7 NP_499869.3 13 - 14 1637 55
GenBank mRNA KM875546.1 - 12 - 13 1565 55

View Full Product Details