Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
col-99
Entrez Gene ID
185112
Gene Name
protein-coding
Gene Aliases
CELE_F29C4.8
Chromosome Location
Chr.IV: 137974 - 147919 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307632.3 NP_001294561.1 3 - 4 370 59
NM_001307633.3 NP_001294562.1 3 - 4 370 59
NM_001307634.4 NP_001294563.1 3 - 4 370 59
NM_001322764.3 NP_001309612.1 3 - 4 370 59
NM_001322765.3 NP_001309608.1 3 - 4 370 59
NM_001322766.3 NP_001309610.1 3 - 4 370 59
NM_001322767.3 NP_001309606.1 3 - 4 370 59
NM_001322768.3 NP_001309611.1 3 - 4 370 59
NM_001322769.3 NP_001309607.1 3 - 4 370 59
NM_001322770.3 NP_001309609.1 3 - 4 370 59
NM_001322771.3 NP_001309605.1 3 - 4 370 59
NM_001322772.3 NP_001309604.1 3 - 4 370 59
NM_001322773.3 NP_001309602.1 3 - 4 370 59
NM_001322774.3 NP_001309603.1 3 - 4 370 59
NM_001322775.2 NP_001309601.1 3 - 4 370 59
NM_067468.7 NP_499869.3 3 - 4 370 59
GenBank mRNA KM875546.1 - 3 - 4 370 59

View Full Product Details