Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Y38C1AA.6
Entrez Gene ID
189640
Gene Name
RING-type domain-containing protein
Gene Aliases
CELE_Y38C1AA.6
Chromosome Location
Chr.IV: 162830 - 169981 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379994.1 NP_001367798.1 4 - 5 1880 69

Gene Symbol
cutl-24
Entrez Gene ID
176831
Gene Name
protein-coding
Gene Aliases
CELE_Y38C1AA.5
Chromosome Location
Chr.IV: 162824 - 169980 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028278.5 NP_001023449.1 4 - 5 1886 69

View Full Product Details